Order Entry
Finland
ContactUsLinkComponent
4-{2-[4-(Trifluoromethyl)piperidino]acetyl}-1H-pyrrole-2-carbaldehyde
4-{2-[4-(Trifluoromethyl)piperidino]acetyl}-1H-pyrrole-2-carbaldehyde
Catalog # APOSPC7998-1G
Supplier:  Apollo Scientific
CAS Number:  
4-{2-[4-(Trifluoromethyl)piperidino]acetyl}-1H-pyrrole-2-carbaldehyde
Catalog # APOSPC7998-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:142…144 °C
 :288.27 g/mol
Storage Temperature:Ambient
CAS Number:1000339-90-1
Formula:C₁₃H₁₅F₃N₂O₂
MDL Number:MFCD09864685