

1,3,5-Tris(trifluoromethyl)benzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:729-81-7
:
Formula:C₉H₃F₉
MDL Number:MFCD00054722
:282.105 g/mol
Boiling Pt:119.6 °C/750mmHg
Melting Pt:9 °C
Density:1.5139 g/cm³ (25 °C)
Flash Pt:43 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07