
1-Butyl-3-methylimidazoliumhexafluorophosphate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:174501-64-5
:
Formula:C₈H₁₅F₆N₂P
MDL Number:MFCD03093295
:284.18 g/mol
Boiling Pt:>340 °C
Melting Pt:44383 °C
Density:1.38 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together