Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2-fluorobenzyl alcohol
5-Bromo-2-fluorobenzyl alcohol
5-Bromo-2-fluorobenzyl alcohol
Irritant
5-Bromo-2-fluorobenzyl alcohol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Bromo-2-fluorobenzyl alcohol
Supplier:
Apollo Scientific
About this item
CAS Number:
99725-13-0
:
Formula:
C₇H₆BrFO
MDL Number:
MFCD00143425
:
205.03
Boiling Pt:
82…87 °C/0.4mmHg
:
GHS07
Storage Temperature:
Ambient