Order Entry
Finland
ContactUsLinkComponent
Tetraethylammonium tetrafluoroborate 97%
Irritant
Tetraethylammonium tetrafluoroborate 97%
Catalog # APOSPC6667-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Tetraethylammonium tetrafluoroborate 97%
Catalog # APOSPC6667-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :217.06 g/mol
Melting Pt:>300 °C
EINECS:207-055-1
 :GHS07
Storage Temperature:Ambient
CAS Number:429-06-1
Formula:C₈H₂₀BF₄N
MDL Number:MFCD00011827