Order Entry
Finland
Orders LinkContactUsLinkComponent
Perfluoroicosane 97%

Perfluoroicosane 97%

Bottle, 1 g
Catalog # APOSPC6004-1G
Irritant
Supplier:   Apollo Scientific
Perfluoroicosane 97%
Catalog # APOSPC6004-1G
Supplier:  Apollo Scientific
CAS Number:  37589-57-4
Perfluoroicosane 97%

Bottle, 1 g

CAS Number:  37589-57-4
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents

Mass spectrometry standard.

  • CAS Number:37589-57-4
  • Formula:C₂₀F₄₂
  • MDL Number:MFCD00000438
  • Molecular Weight:1038.15 g/mol
  • Boiling Pt:320 °C
  • Melting Pt:162…165 °C
  • Storage Temperature:Ambient
  • EINECS:253-557-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.