Order Entry
Finland
ContactUsLinkComponent
4-Bromo-2-fluoro-5-methoxyphenylboronic acid pinacol ester 96%
Irritant
4-Bromo-2-fluoro-5-methoxyphenylboronic acid pinacol ester 96%
Catalog # APOSPC5753-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromo-2-fluoro-5-methoxyphenylboronic acid pinacol ester 96%
Catalog # APOSPC5753-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :330.99 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1150271-71-8
Formula:C₁₃H₁₇BBrFO₃
MDL Number:MFCD11855986