Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(2-Methoxyethylamino)-3-nitrobenzotrifluoride
4-(2-Methoxyethylamino)-3-nitrobenzotrifluoride
4-(2-Methoxyethylamino)-3-nitrobenzotrifluoride
4-(2-Methoxyethylamino)-3-nitrobenzotrifluoride
Supplier:
Apollo Scientific
Certificates
Expand image
4-(2-Methoxyethylamino)-3-nitrobenzotrifluoride
Supplier:
Apollo Scientific
About this item
CAS Number:
161605-63-6
:
Formula:
C₁₀H₁₁F₃N₂O₃
MDL Number:
MFCD00798374
Storage Temperature:
Ambient