0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
2-Nitro-6-(trifluoromethyl)anisole
2-Nitro-6-(trifluoromethyl)anisole
2-Nitro-6-(trifluoromethyl)anisole
Corrosive
Irritant
2-Nitro-6-(trifluoromethyl)anisole
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
2-Nitro-6-(trifluoromethyl)anisole
Supplier:
Apollo Scientific
About this item
CAS Number:
627531-40-2
:
Formula:
C₈H₆F₃NO₃
MDL Number:
MFCD08532488
Storage Temperature:
Ambient
UN:
1760
ADR:
8,II
:
GHS05
:
GHS07