Order Entry
Finland
ContactUsLinkComponent
2-Methyl-4-nitrobenzotrifluoride 97%
2-Methyl-4-nitrobenzotrifluoride 97%
Catalog # APOSPC520626-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-4-nitrobenzotrifluoride 97%
Catalog # APOSPC520626-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :205.14 g/mol
Storage Temperature:Ambient
CAS Number:1960-52-7
Formula:C₈H₆F₃NO₂
MDL Number:MFCD11226539