
5-Amino-2-chloro-4-nitrobenzotrifluoride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:167415-22-7
:
Formula:C₇H₄ClF₃N₂O₂
MDL Number:MFCD03094264
:240.5700 g/mol
Melting Pt:123…125 °C
:GHS07
Storage Temperature:Ambient