Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid 98%
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid 98%
Irritant
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
481681-02-1
:
Formula:
C₇H₈BFO₃
MDL Number:
MFCD08235075
:
169.95 g/mol
Melting Pt:
124…126 °C
:
GHS07
Storage Temperature:
Ambient