Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Difluoro-4-nitrobenzonitrile
3,5-Difluoro-4-nitrobenzonitrile 98%
3,5-Difluoro-4-nitrobenzonitrile 98%
Irritant
3,5-Difluoro-4-nitrobenzonitrile 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Difluoro-4-nitrobenzonitrile 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1123172-88-2
:
Formula:
C7H2F2N2O2
MDL Number:
MFCD13193247
:
184.0998 g/mol
Storage Temperature:
Refrigerator
UN:
2811
ADR:
6.1,III
:
GHS07