Order Entry
Finland
ContactUsLinkComponent
N-(Hydroxymethyl)-2,2,2-trifluoroacetamide
Irritant
N-(Hydroxymethyl)-2,2,2-trifluoroacetamide
Catalog # APOSPC49320-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-(Hydroxymethyl)-2,2,2-trifluoroacetamide
Catalog # APOSPC49320-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :143.06 g/mol
Melting Pt:99…103 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:50667-69-1
Formula:C₃H₄F₃NO₂
MDL Number:MFCD00014416