

Phenylsulphur pentafluoride 98+%
Catalog # APOSPC49103-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:148 °C
Density:1.496 g/cm³ (25 °C)
Flash Pt:43.3 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:2557-81-5
Formula:C₆H₅F₅S
MDL Number:MFCD09263740