(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride 95+%
Catalog # APOSPC48541-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:193.63 g/mol
Storage Temperature:Ambient
CAS Number:844647-37-6
Formula:C₈H₁₀ClF₂N
MDL Number:MFCD11101244