Order Entry
Finland
ContactUsLinkComponent
2,2,3,3,4,4-Hexafluoro-1,5-pentanediol 98%
Irritant
2,2,3,3,4,4-Hexafluoro-1,5-pentanediol 98%
Catalog # APOSPC4770-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,2,3,3,4,4-Hexafluoro-1,5-pentanediol 98%
Catalog # APOSPC4770-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :212.09 g/mol
Boiling Pt:110…112 °C/10mmHg
Melting Pt:76…80 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:206-819-1
 :GHS07
CAS Number:376-90-9
Formula:C₅H₆F₆O₂
MDL Number:MFCD00042434