Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluoro-4-nitrobenzoic acid
3-Fluoro-4-nitrobenzoic acid 97%
3-Fluoro-4-nitrobenzoic acid 97%
Irritant
3-Fluoro-4-nitrobenzoic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Fluoro-4-nitrobenzoic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
403-21-4
:
Formula:
C₇H₄FNO₄
MDL Number:
MFCD01862092
:
185.11 g/mol
Melting Pt:
173…176 °C
Flash Pt:
>110 °C
:
GHS07
Storage Temperature:
Ambient