
Fomblin® Y L-VAC 14/6
Catalog # APOSPC4382-1KG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Density:1.9 g/cm³ (25 °C)
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
CAS Number:69991-67-9
MDL Number:MFCD00163696