4-(4-Bromo-2-fluorobenzyl)morpholine
Catalog # APOSPC430519-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:274.133 g/mol
Storage Temperature:Ambient
CAS Number:338454-98-1
Formula:C₁₁H₁₃BrFNO
MDL Number:MFCD12022322