2-Fluorophenyl isocyanate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16744-98-2
:
Formula:C₇H₄FNO
MDL Number:MFCD00001996
Boiling Pt:56…58 °C/10mmHg
Density:1.222 g/cm³ (25 °C)
Flash Pt:50 °C
Storage Temperature:Refrigerator
EINECS:240-804-0
UN:3080
ADR:6.1+3,II
:137.11 g/mol
:GHS02
:GHS06
:GHS08