Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2-fluorobenzyl chloride
4-Bromo-2-fluorobenzyl chloride 97%
4-Bromo-2-fluorobenzyl chloride 97%
4-Bromo-2-fluorobenzyl chloride 97%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Bromo-2-fluorobenzyl chloride 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
85510-82-3
:
Formula:
C₇H₅BrClF
MDL Number:
MFCD02094036
:
223.47 g/mol
Storage Temperature:
Ambient