
2-Fluoro-3-methylbenzyl bromide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:151412-12-3
:
Formula:C₈H₈BrF
MDL Number:MFCD00042470
Boiling Pt:168 °C/15mmHg
UN:3265
ADR:8,II
:203.05 g/mol
:GHS05
Storage Temperature:Ambient