Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-4-nitrobenzotrifluoride
3-Bromo-4-nitrobenzotrifluoride 97%
3-Bromo-4-nitrobenzotrifluoride 97%
APOSPC3153-5G
:
Apollo Scientific
:
3-Bromo-4-nitrobenzotrifluoride 97%
APOSPC3153-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
270.01 g/mol
:
Ambient
:
132839-58-8
:
C₇H₃BrF₃NO₂
:
MFCD09998160