Order Entry
Finland
ContactUsLinkComponent
4-Methoxy-2-(trifluoromethyl)phenylboronic acid 98%
Irritant
4-Methoxy-2-(trifluoromethyl)phenylboronic acid 98%
Catalog # APOSPC3068-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Methoxy-2-(trifluoromethyl)phenylboronic acid 98%
Catalog # APOSPC3068-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :219.95 g/mol
Melting Pt:168…172 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:313546-16-6
Formula:C₈H₈BF₃O₃
MDL Number:MFCD03095346