Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Difluorobenzyl alcohol
3,5-Difluorobenzyl alcohol 98%
3,5-Difluorobenzyl alcohol 98%
:
Apollo Scientific
3,5-Difluorobenzyl alcohol 98%
:
Apollo Scientific
:
79538-20-8
:
:
C₇H₆F₂O
:
MFCD00010313
:
144.12 g/mol
:
98…99 °C/9mmHg
:
1.276 g/cm³ (25 °C)
:
94 °C
:
GHS07
:
Ambient