Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dibromo-6-fluoroaniline
2,4-Dibromo-6-fluoroaniline 97%
2,4-Dibromo-6-fluoroaniline 97%
2,4-Dibromo-6-fluoroaniline 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Dibromo-6-fluoroaniline 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
141474-37-5
:
Formula:
C₆H₄Br₂FN
MDL Number:
MFCD00042230
Melting Pt:
62…64 °C
:
268.91 g/mol
:
GHS07
Storage Temperature:
Ambient