Order Entry
Finland
ContactUsLinkComponent
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 98%
 
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 98%
  APOSPC201120-5G
 :  
 
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 98%
  APOSPC201120-5G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

 :
 :196.17 g/mol
 :GHS07
 :Ambient
 :133261-33-3
 :C₈H₁₁F₃O₂
 :MFCD09037363