Order Entry
Finland
ContactUsLinkComponent
Chloropentafluorobenzene 99%
Toxic
Chloropentafluorobenzene 99%
Catalog # APOSPC2010-5G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
Chloropentafluorobenzene 99%
Catalog # APOSPC2010-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :202.51 g/mol
Boiling Pt:122…123 °C/750mmHg
Density:1.568 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:206-450-6
UN:2810
ADR:6.1,III
 :GHS06
CAS Number:344-07-0
Formula:C₆ClF₅
MDL Number:MFCD00000534