
2-[(tert-Butoxycarbonyl)amino]-3-(trifluoromethoxy)benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:561304-40-3
:
Formula:C₁₃H₁₄F₃NO₅
MDL Number:MFCD09800581
:321.26 g/mol
Melting Pt:135…136 °C
:GHS07
Storage Temperature:Ambient