Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-6-fluoro-2-(trifluoromethyl)quinoline
4-Chloro-6-fluoro-2-(trifluoromethyl)quinoline 97%
4-Chloro-6-fluoro-2-(trifluoromethyl)quinoline 97%
Irritant
4-Chloro-6-fluoro-2-(trifluoromethyl)quinoline 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Chloro-6-fluoro-2-(trifluoromethyl)quinoline 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
59611-55-1
:
Formula:
C₁₀H₄ClF₄N
MDL Number:
MFCD00276579
Melting Pt:
78…81 °C
:
GHS07
Storage Temperature:
Ambient