Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Bromo-3,4,5-trifluorobenzene
1-Bromo-3,4,5-trifluorobenzene 99%
1-Bromo-3,4,5-trifluorobenzene 99%
:
Apollo Scientific
1-Bromo-3,4,5-trifluorobenzene 99%
:
Apollo Scientific
:
138526-69-9
:
:
C₆H₂BrF₃
:
MFCD00070738
:
210.98 g/mol
:
47…49 °C/60mmHg
:
< –20 °C
:
1.767 g/cm³ (25 °C)
:
45 °C
:
418-480-9
:
1993
:
3,III
:
GHS02
:
GHS05
:
GHS08
:
GHS09
:
Ambient