

α,4-Dibromo-2-fluorotoluene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:76283-09-5
:
Formula:C₇H₅Br₂F
MDL Number:MFCD00055467
:267.92 g/mol
Boiling Pt:126 °C/19mmHg
Melting Pt:30 °C
Storage Temperature:Ambient
EINECS:278-412-7
UN:2923
ADR:8+6.1,III
:GHS05
:GHS06