Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluoro-2-nitrobenzonitrile
3-Fluoro-2-nitrobenzonitrile 96%
3-Fluoro-2-nitrobenzonitrile 96%
Toxic
3-Fluoro-2-nitrobenzonitrile 96%
Supplier:
Apollo Scientific
Certificates
Expand image
Toxic
3-Fluoro-2-nitrobenzonitrile 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
1000339-52-5
:
Formula:
C₇H₃FN₂O₂
MDL Number:
MFCD09864664
:
166.1100 g/mol
Storage Temperature:
Ambient
UN:
3439
ADR:
6.1,III
:
GHS06