Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium (2-methoxyphenyl)trifluoroborate
Potassium (2-methoxyphenyl)trifluoroborate 98%
Potassium (2-methoxyphenyl)trifluoroborate 98%
Irritant
Potassium (2-methoxyphenyl)trifluoroborate 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Potassium (2-methoxyphenyl)trifluoroborate 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
236388-46-8
:
Formula:
C₇H₇BF₃KO
MDL Number:
MFCD04115756
:
214.04 g/mol
Melting Pt:
282…286 °C
:
GHS07
Storage Temperature:
Ambient