Order Entry
Finland
ContactUsLinkComponent
1-Fluoro-3,3-dimethyl-1,2-benziodoxole 98%
Irritant
1-Fluoro-3,3-dimethyl-1,2-benziodoxole 98%
Catalog # APOSPC10688-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Fluoro-3,3-dimethyl-1,2-benziodoxole 98%
Catalog # APOSPC10688-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :280.08 g/mol
Melting Pt:88 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1391728-13-4
Formula:C₉H₁₀FIO
MDL Number:MFCD28397076