Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluoro-5-bromobenzyl bromide
2-Fluoro-5-bromobenzyl bromide 97%
2-Fluoro-5-bromobenzyl bromide 97%
2-Fluoro-5-bromobenzyl bromide 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Fluoro-5-bromobenzyl bromide 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
99725-12-9
:
Formula:
C₇H₅Br₂F
MDL Number:
MFCD09038457
:
267.923 g/mol
Storage Temperature:
Ambient
UN:
3261
ADR:
8,III
:
GHS05