Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluoro-4-(trifluoromethoxy)benzaldehyde
3-Fluoro-4-(trifluoromethoxy)benzaldehyde 98%
3-Fluoro-4-(trifluoromethoxy)benzaldehyde 98%
:
Apollo Scientific
3-Fluoro-4-(trifluoromethoxy)benzaldehyde 98%
:
Apollo Scientific
:
473917-15-6
:
:
C₈H₄F₄O₂
:
MFCD00236275
:
208.11 g/mol
:
GHS07
:
Ambient