Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pentafluorophenyl 2-fluorobenzenesulphonate
Pentafluorophenyl-2-fluorobenzenesulphonate
Pentafluorophenyl-2-fluorobenzenesulphonate
Pentafluorophenyl-2-fluorobenzenesulphonate
Supplier:
Apollo Scientific
Certificates
Expand image
Pentafluorophenyl-2-fluorobenzenesulphonate
Supplier:
Apollo Scientific
About this item
CAS Number:
885950-35-6
:
Formula:
C₁₂H₄F₆O₃S
MDL Number:
MFCD05975126
Melting Pt:
104…105 °C
Storage Temperature:
Ambient