Order Entry
Finland
ContactUsLinkComponent
4-Amino-2-chloro-5-nitrobenzotrifluoride 97%
Irritant
4-Amino-2-chloro-5-nitrobenzotrifluoride 97%
Catalog # APOSPC0912-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Amino-2-chloro-5-nitrobenzotrifluoride 97%
Catalog # APOSPC0912-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :240.5700 g/mol
Melting Pt:113…114 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:35375-74-7
Formula:C₇H₄ClF₃N₂O₂
MDL Number:MFCD01312277