Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Difluorophenylboronic acid
3,4-Difluorophenylboronic acid 98%
3,4-Difluorophenylboronic acid 98%
:
Apollo Scientific
3,4-Difluorophenylboronic acid 98%
:
Apollo Scientific
:
168267-41-2
:
:
C₆H₅BF₂O₂
:
MFCD00807405
:
157.91 g/mol
:
305…310 °C
:
GHS07
:
Ambient