Order Entry
Finland
ContactUsLinkComponent
 
Flammable
Irritant
α,α,α,2,4,6-Hexafluorotoluene
Catalog # APOSPC0514-1G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
α,α,α,2,4,6-Hexafluorotoluene
Catalog # APOSPC0514-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.08 g/mol
Boiling Pt:105…107 °C
Density:1.475 g/cm³ (25 °C)
Flash Pt:51 °C
UN:1993
ADR:3,III
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:122030-04-0
Formula:C₇H₂F₆
MDL Number:MFCD00190109