Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Fluoro-2-methylbenzyl alcohol
5-Fluoro-2-methylbenzyl alcohol
5-Fluoro-2-methylbenzyl alcohol
Irritant
5-Fluoro-2-methylbenzyl alcohol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Fluoro-2-methylbenzyl alcohol
Supplier:
Apollo Scientific
About this item
CAS Number:
22062-54-0
:
Formula:
C₈H₉FO
MDL Number:
MFCD01631527
:
140.16 g/mol
Melting Pt:
36…40 °C
:
GHS07
Storage Temperature:
Ambient