

α-Bromo-2-chloro-5-fluorotoluene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:81778-09-8
:
Formula:C₇H₅BrClF
MDL Number:MFCD01631419
Boiling Pt:65 °C/2mmHg
Density:1.654 g/cm³ (25 °C)
Flash Pt:17.8 °C
UN:2924
ADR:3+8,II
:223.47 g/mol
:GHS02
:GHS05
Storage Temperature:Refrigerator