Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%
Irritant
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
2199-44-2
:
Formula:
C₉H₁₃NO₂
MDL Number:
MFCD00010645
Melting Pt:
125…126 °C
:
167.21 g/mol
:
GHS07
Storage Temperature:
Ambient