Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(Allyloxycarbonyloxy)succinimide
N-(Allyloxycarbonyloxy)succinimide 98%
N-(Allyloxycarbonyloxy)succinimide 98%
:
Apollo Scientific
N-(Allyloxycarbonyloxy)succinimide 98%
:
Apollo Scientific
:
135544-68-2
:
:
C₈H₉NO₅
:
MFCD02684388
:
199.16 g/mol
:
GHS07
:
Freezer