
4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) 95+%
Catalog # APOSOR960249-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pakkaustyyppi:Bottle
- Pak.:25 g
- Seq:3308
Specifications
About this item
:
:144.17 g/mol
:GHS07
Storage Temperature:Huoneenlämpö
CAS Number:17419-81-7
Formula:C₇H₁₂O₃
MDL Number:MFCD00077974