3,3-Dimethyl-3,4-dihydroquinoxalin-2(1H)-one 98%
Catalog # APOSOR952582-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:176.22 g/mol
Storage Temperature:Ambient
CAS Number:80636-30-2
Formula:C₁₀H₁₂N₂O
MDL Number:MFCD00186947