0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
4-Iodo-L-phenylalanine
4-Iodo-L-phenylalanine
4-Iodo-L-phenylalanine
Irritant
4-Iodo-L-phenylalanine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Iodo-L-phenylalanine
Supplier:
Apollo Scientific
About this item
CAS Number:
24250-85-9
:
Formula:
C₉H₁₀INO₂
MDL Number:
MFCD00002602
:
291.1 g/mol
:
GHS07
Storage Temperature:
Freezer