Ethylene glycol mono-m-tolyl ether 98%
Catalog # APOSOR937919-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:152.19 g/mol
Storage Temperature:Ambient
CAS Number:13605-19-1
Formula:C₉H₁₂O₂
MDL Number:MFCD00014035